Learn More
Integra Biosciences Corp 12 CHANNEL EVOLVE
SDP
Catalog No. NC1672018
Encompass
The EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.
Catalog No. NC1672018 Supplier Integra Biosciences Corp Supplier No. 3032
The EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.