Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well, 19 Gauge, SS Tubes, 63.5mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC1621258
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 63.5mm Long, 9mm Center Spacing, Polycarbonate, includes VP 187S-1
Catalog No. NC1621258 Supplier V&P Scientific Inc Supplier No. VP 187PC3S
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 63.5mm Long, 9mm Center Spacing, Polycarbonate, includes VP 187S-1