Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR, 19 Gauge, SS Tubes, 16mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC1651183
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR Plate, 19 Gauge, Stainless Steel Tubes, 16mm Long, 9 mm Center Spacing, Polycarbonate, includes VP 187S-1
Catalog No. NC1651183 Supplier V&P Scientific Inc Supplier No. VP 187PC2S
ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR Plate, 19 Gauge, Stainless Steel Tubes, 16mm Long, 9 mm Center Spacing, Polycarbonate, includes VP 187S-1