Learn More
V&P Scientific Inc ASPIRATION WAND, 12 Channel, 96 Deep Well, 20 Gauge Tubes, 46mm Long, 9mm Spacing, Polypropylene
Catalog No. NC0979260
Encompass
ASPIRATION WAND, 12 Channel, 96 Well, Deep Well Plate, 20 Gauge, Stainless Steel Tubes, 46mm Long, 9mm Center Spacing, Polypropylene
Catalog No. NC0979260 Supplier V&P Scientific Inc Supplier No. VP 185BP
ASPIRATION WAND, 12 Channel, 96 Well, Deep Well Plate, 20 Gauge, Stainless Steel Tubes, 46mm Long, 9mm Center Spacing, Polypropylene