Learn More
Research Products International Corp Twelve Channel Reagent Reservoir with Lid, Non-Sterile, 10 per Package
SDP
Catalog No. 50476519
Encompass
Ideal reagent reservoir for performing serial dilutions or pipetting different reagents with a multi-channel pipettor. Each clearly numbered chamber holds up to 3ml of reagent and has a ''V'' shaped bottom. Made from chemical resistant polypropylene that can also be autoclaved. Each reservoir is individually wrapped to insure cleanliness and supplied with a clear lid.
Catalog No. 50-476-519 Supplier Research Products International Corp Supplier No. 246270
Add to Cart
Add to Cart
Ideal reagent reservoir for performing serial dilutions or pipetting different reagents with a multi-channel pipettor. Each clearly numbered chamber holds up to 3ml of reagent and has a ''V'' shaped bottom. Made from chemical resistant polypropylene that can also be autoclaved. Each reservoir is individually wrapped to insure cleanliness and supplied with a clear lid.