Learn More
USA Scientific Inc 12-channel reservoir for automation
Catalog No. NC9056368
Encompass
Twelve well polypropylene reservoir, automation compatible footprint, 128 mm L x 86 mm W x 44 mm H. 5/pack
Catalog No. NC9056368 Supplier USA Scientific Inc Supplier No. 10618150
Twelve well polypropylene reservoir, automation compatible footprint, 128 mm L x 86 mm W x 44 mm H. 5/pack