Learn More
Andwin Scientific 12-CHANNEL RESERVOIR NATURAL
SDP
Catalog No. 50819460
Encompass_Preferred
This reservoir features 12 channels that each have the capacity of up to 22 mL. Each section has eight dimples across the base, which allows maximum recovery with multi-channel devices. There are also bold numeric markings for each section that permit easy identification. This product is autoclavable. It comes in a pack of 5 128 mm x 86 mm x 44 mm reservoirs.
Catalog No. 50-819-460 Supplier Andwin Scientific Supplier No. 10618150
This reservoir features 12 channels that each have the capacity of up to 22 mL. Each section has eight dimples across the base, which allows maximum recovery with multi-channel devices. There are also bold numeric markings for each section that permit easy identification. This product is autoclavable. It comes in a pack of 5 128 mm x 86 mm x 44 mm reservoirs.