Learn More
Thomas Scientific 12-Channel Pipet Basins, Sterile
SDP
Catalog No. NC0192531
Encompass_Preferred
Virgin polypropylene material Numbered 5 mL channels Wide temperature service range Designed for serial dilutions or pipetting different reagents using up to a 12-channel pipettor. Each of the 12 chambers holds up to 5mL and is “V” shaped to facilitate complete sample withdrawal. Each chamber is numbered to ensure accurate sample reference. Made from virgin polypropylene, the basin offers excellent chemical resistance and can withstand temperatures from -80Ý to +121ÝC.
Catalog No. NC0192531 Supplier Thomas Scientific Supplier No. 1231D25
Virgin polypropylene material Numbered 5 mL channels Wide temperature service range Designed for serial dilutions or pipetting different reagents using up to a 12-channel pipettor. Each of the 12 chambers holds up to 5mL and is “V” shaped to facilitate complete sample withdrawal. Each chamber is numbered to ensure accurate sample reference. Made from virgin polypropylene, the basin offers excellent chemical resistance and can withstand temperatures from -80Ý to +121ÝC.