Learn More
Arcticwhite LLC 12 Column Partitioned Reservoir, Pyramid Bottom, Non-sterilized, 12 Columns, Polypropylene (PP), Pyramid, Partitioned, 21 ML/Column, 25 Reagent Reservoirs per Case
SDP
Catalog No. NC1645930
Encompass
12 Column Partitioned Reservoir, Pyramid Bottom, Non-sterilized, 12 Columns, Polypropylene (PP), Pyramid, Partitioned, 21 ML/Column, 25 Reagent Reservoirs per Case. This 12-column reagent reservoir has partitioned channels and a pyramid bottom for use with automated liquid handlers and multichannel pipettes. Format - 12 columns. Bottom - pyramid, partitioned. Capacity - 21 mL/Column. Material - polypropylene (PP). Packaging - case of 25. Supplied by Arctic White as 25 reagent reservoirs per case.
Catalog No. NC1645930 Supplier Arcticwhite LLC Supplier No. AWLSS30019
12 Column Partitioned Reservoir, Pyramid Bottom, Non-sterilized, 12 Columns, Polypropylene (PP), Pyramid, Partitioned, 21 ML/Column, 25 Reagent Reservoirs per Case. This 12-column reagent reservoir has partitioned channels and a pyramid bottom for use with automated liquid handlers and multichannel pipettes. Format - 12 columns. Bottom - pyramid, partitioned. Capacity - 21 mL/Column. Material - polypropylene (PP). Packaging - case of 25. Supplied by Arctic White as 25 reagent reservoirs per case.