Learn More
Thomas Scientific AxyVoir, Pre-Sterlized Low Profile Multi Well Individually Wrapped, 12 Bottom Reservoir, 25/cs
SDP
Catalog No. NC0863012
Encompass
Polypropylene reagent vessels. Reservoirs are made of clear virgin polypropylene for excellent chemical resistance. Features an SBS footprint. Various formats for any application.
Catalog No. NC0863012 Supplier Thomas Scientific Supplier No. 1223W08
Polypropylene reagent vessels. Reservoirs are made of clear virgin polypropylene for excellent chemical resistance. Features an SBS footprint. Various formats for any application.