Learn More
Med Vet International 12" Cotton Removal Tweezers, J1118
Catalog No. 5000585742
Encompass_Preferred
These 12" tweezers make it easy to remove cotton or pills from deep bottles. Stainless steel.
Catalog No. 50-005-85742 Supplier Med Vet International Supplier No. JOR1118
These 12" tweezers make it easy to remove cotton or pills from deep bottles. Stainless steel.