Learn More
Bio Rad Laboratories 12% Criterion TGX Precast Midi Protein Gel, 18 well, 30 uL
Catalog No. NC1316309
Encompass
Pkg of 1, 12% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells.
Catalog No. NC1316309 Supplier Bio Rad Laboratories Supplier No. 5671044
Pkg of 1, 12% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells.