Learn More
Medchemexpress LLC 12-crown-4 | 294-93-9 | MFCD00005103 | 98.0% | 176.21 | C8H16O4 | 250 MG
SDP
Catalog No. 5000302382
Encompass_Preferred
12-Crown-4 is a crown ether used as a phase-transfer catalyst and a lithium-complexing agent for laboratory and research applications. It is supplied as a high-purity reagent (98.0%) with formula C8H16O4 and molecular weight 176.21 g·mol-1, available in small pack sizes for synthesis and analytical workflows.
  • High purity (98.0%) suitable for research applications.
  • Effective phase-transfer catalyst for organic synthesis.
  • Forms selective complexes with lithium cations.
  • Liquid at ambient conditions for easy dosing and handling.
  • Stable when stored under recommended temperatures.
Catalog No. 50-003-02382 Supplier Medchemexpress LLC Supplier No. 250MGHYW016693
12-Crown-4 is a crown ether used as a phase-transfer catalyst and a lithium-complexing agent for laboratory and research applications. It is supplied as a high-purity reagent (98.0%) with formula C8H16O4 and molecular weight 176.21 g·mol-1, available in small pack sizes for synthesis and analytical workflows.
  • High purity (98.0%) suitable for research applications.
  • Effective phase-transfer catalyst for organic synthesis.
  • Forms selective complexes with lithium cations.
  • Liquid at ambient conditions for easy dosing and handling.
  • Stable when stored under recommended temperatures.