Learn More
Medchemexpress LLC 12-crown-4 | 294-93-9 | MFCD00005103 | 98.0% | 176.21 g·mol⁻¹ | C8H16O4 | 500 MG
SDP
Catalog No. 5000299383
Encompass_Preferred
12-crown-4 is a crown ether (1,4,7,10-tetraoxacyclododecane) that selectively complexes small alkali metal cations, especially lithium. It is supplied as a high-purity reagent for use as a phase-transfer catalyst, ionophore, and ligand in synthetic and analytical chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Selective complexation of lithium and other small alkali metal cations.
  • Facilitates phase-transfer catalysis between immiscible solvents.
  • Useful as an ionophore in coordination chemistry and sensing studies.
  • Available in small laboratory pack sizes for bench-scale work.
Catalog No. 50-002-99383 Supplier Medchemexpress LLC Supplier No. HYW016693500MG
12-crown-4 is a crown ether (1,4,7,10-tetraoxacyclododecane) that selectively complexes small alkali metal cations, especially lithium. It is supplied as a high-purity reagent for use as a phase-transfer catalyst, ionophore, and ligand in synthetic and analytical chemistry.
  • High purity suitable for analytical and synthetic applications.
  • Selective complexation of lithium and other small alkali metal cations.
  • Facilitates phase-transfer catalysis between immiscible solvents.
  • Useful as an ionophore in coordination chemistry and sensing studies.
  • Available in small laboratory pack sizes for bench-scale work.