Learn More
Selleck Chemical LLC 12-Crown-4 E4834-1G
SDP
Catalog No. 5000041741
Encompass_Preferred
12-crown-4(EOCT Ethylene oxide cyclic tetramer) is an effective phase-transfer catalyst and complexing agent for lithium facilitating the dissolution of lithium bis(trifluoromethanesulfonyl)imide (LiTFSI) without requiring acetonitrile It selectively binds to lithium ions and forms stable complexes significantly improving electrolyte conductivity in lithium batteries and positively influencing ion mobility and overall battery performance
Catalog No. 50-000-41741 Supplier Selleck Chemical LLC Supplier No. E48341G
12-crown-4(EOCT Ethylene oxide cyclic tetramer) is an effective phase-transfer catalyst and complexing agent for lithium facilitating the dissolution of lithium bis(trifluoromethanesulfonyl)imide (LiTFSI) without requiring acetonitrile It selectively binds to lithium ions and forms stable complexes significantly improving electrolyte conductivity in lithium batteries and positively influencing ion mobility and overall battery performance