Learn More
Medchemexpress LLC 12-((cyclohexylcarbamoyl)amino)dodecanoic acid | 479413-68-8 | 99.8% | 10 MG
SDP
Catalog No. 5000232577
Encompass_Preferred
CUDA is a research-grade small-molecule inhibitor of soluble epoxide hydrolase (sEH) used in biochemical and cellular studies to probe sEH and PPAR-alpha pathways. It is supplied as a crystalline solid (CAS 479413-68-8) with molecular formula C19H36N2O3 and molecular weight 340.50 g/mol.
  • Potent sEH inhibitor with IC50 = 11.1 nM (mouse) and 112 nM (human).
  • High purity by HPLC (approximately 99.8%).
  • Supplied in small solid quantities suitable for assay development and cellular studies.
  • Soluble in common organic solvents for stock solution preparation.
Catalog No. 50-002-32577 Supplier Medchemexpress LLC Supplier No. HY12153810MG
CUDA is a research-grade small-molecule inhibitor of soluble epoxide hydrolase (sEH) used in biochemical and cellular studies to probe sEH and PPAR-alpha pathways. It is supplied as a crystalline solid (CAS 479413-68-8) with molecular formula C19H36N2O3 and molecular weight 340.50 g/mol.
  • Potent sEH inhibitor with IC50 = 11.1 nM (mouse) and 112 nM (human).
  • High purity by HPLC (approximately 99.8%).
  • Supplied in small solid quantities suitable for assay development and cellular studies.
  • Soluble in common organic solvents for stock solution preparation.