Learn More
Enzo Life Sciences 12-Deoxyphorbol 13-acetate (1mg). CAS: 60857-08-1
SDP
Supplier:  Enzo Life Sciences BMLPE1870001
Encompass_Preferred
A non-tumor promoting phorbol ester, which acts as a PKC partial agonist. Protects T-lymphoblastoid cells from the killing effects of HIV-1. Upregulates latent HIV-1 provirus expression and inhibits HIV infection and viral spread at the entry/fusion step of viral life cycle. Does not induce tumor formation. Alternative name: Prostratin. Purity: ≥99% (HPLC, TLC). Solubility: Soluble in 100% ethanol (25mg/ml) or DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1408