Learn More
MSC Tesa Brown and Sharpe Dial Caliper, 0-12 in., Stainless Steel, 1 ea.
Supplier:  MSC 63739577
Encompass
This dial caliper is a precision measurement tool designed for professional applications requiring high accuracy and reliability. Constructed from durable stainless steel, it features a patented shock-proof and anti-backlash gear mechanism to ensure performance in demanding environments. This versatile instrument provides 4-way measurement capabilities, including inside, outside, depth, and step dimensions, with a measurement range of 0 to 12 in.
  • Measurement range of 0 to 12 in with 0.001 in graduations
  • Four-way measurement for inside, outside, depth, and step dimensions
  • Shock-proof and anti-backlash design protects the internal gear mechanism
  • Slide lock allows for readings in restricted locations
  • Thumb wheel for smooth jaw adjustment
  • Durable stainless steel construction with a satin chrome finish
  • Includes protective case and a certificate of inspection
Catalog No. 50-005-13874