Learn More
Sigma Aldrich Fine Chemicals Biosciences 12-Dichloroethane Pharmace
Catalog No. 502897443
Encompass
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Catalog No. 50-289-7443 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHR21083X1.2ML
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards