Learn More
Abcam 1,2-Dipalmitoyl-sn-glycero-3-phosphorylglycerol sodium salt (DPPG), Phosphorylglycerol, 100MG
Catalog No. 502706054
Encompass_Preferred
MW 745 Da, Purity >98%. Phosphorylglycerol acylated with palmitic acid. For use in lipid bilayer studies and biological systems. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6054 Supplier Abcam Supplier No. AB143953100MG
MW 745 Da, Purity >98%. Phosphorylglycerol acylated with palmitic acid. For use in lipid bilayer studies and biological systems. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement