Learn More
Medchemexpress LLC 12-dipalmitoyl-sn-glycero-3-ps-d62 sodium | 327178-94-9 | MFCD00674412 | 99.0% | 820.33 | C38H11D62NNaO10P | 5 MG
SDP
Catalog No. 5000221379
Encompass_Preferred
12-Dipalmitoyl-sn-glycero-3-PS-d62 sodium is a deuterium-labeled phosphatidylserine (sodium salt) provided as a stable isotope standard for analytical and lipid research. It is designed for use in mass spectrometry, lipidomics, membrane studies, and method development where isotopic labeling or internal standards are required.
  • Deuterium-labeled (d62) for accurate mass spectrometry quantitation.
  • High purity (99.0%) suitable for analytical workflows.
  • Sodium salt form compatible with common lipid extraction and reconstitution protocols.
  • Supplied in milligram quantities for isotopic standards and tracer experiments.
  • Useful for lipidomics, membrane biophysics, and method development applications.
Catalog No. 50-002-21379 Supplier Medchemexpress LLC Supplier No. HY145539S5MG
12-Dipalmitoyl-sn-glycero-3-PS-d62 sodium is a deuterium-labeled phosphatidylserine (sodium salt) provided as a stable isotope standard for analytical and lipid research. It is designed for use in mass spectrometry, lipidomics, membrane studies, and method development where isotopic labeling or internal standards are required.
  • Deuterium-labeled (d62) for accurate mass spectrometry quantitation.
  • High purity (99.0%) suitable for analytical workflows.
  • Sodium salt form compatible with common lipid extraction and reconstitution protocols.
  • Supplied in milligram quantities for isotopic standards and tracer experiments.
  • Useful for lipidomics, membrane biophysics, and method development applications.