Learn More
Abcam 1,2-Distearoyl-sn-glycero-3-phosphorylglycerol sodium salt (DSPG), Phosphorylglycerol, 100MG
Catalog No. 502706053
Encompass_Preferred
MW 801.1 Da, Purity >98%. Phosphorylglycerol acylated with stearic acid. Enhances respiratory mucus clearance in cystic fibrosis. For use in phospholipid studies and biological systems. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6053 Supplier Abcam Supplier No. AB143954100MG
MW 801.1 Da, Purity >98%. Phosphorylglycerol acylated with stearic acid. Enhances respiratory mucus clearance in cystic fibrosis. For use in phospholipid studies and biological systems. See similar compounds .
The product is subject to the following: Abcam Restricted Use Statement