Learn More
Cayman Chemical 12-doxyl Stearic Acid, Purity: ≥95%, CAS: 29545-47-9, 10 mg
SDP
Catalog No. 502982826
Encompass_Preferred
A form of stearic acid that contains a DOXYL group, creating a hydrophobic spin label
Catalog No. 50-298-2826 Supplier Cayman Chemical Supplier No. 1925910 MG
A form of stearic acid that contains a DOXYL group, creating a hydrophobic spin label