Learn More
BTNX Inc Rapid Response™ 12-Panel Multi-Drug Test Panel
Catalog No. NC1901475
Encompass_Preferred
The Rapid Response™ Multi-Drug Test Panel is a rapid, one step, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine. Test up to 12 drug parameters simultaneously and in just 5 minutes, test results can be observed through the visual interpretation of colored lines.This test is a convenient one-step test. Each test panel is individually sealed in a pouch and comes with a cap that can be used to cover the absorbent strips after dipping in urine sample for a mess-free workspace.
This item is not returnable. View return policy
Catalog No. NC1901475 Supplier BTNX Inc Supplier No. D12.21P2925
The Rapid Response™ Multi-Drug Test Panel is a rapid, one step, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine. Test up to 12 drug parameters simultaneously and in just 5 minutes, test results can be observed through the visual interpretation of colored lines.This test is a convenient one-step test. Each test panel is individually sealed in a pouch and comes with a cap that can be used to cover the absorbent strips after dipping in urine sample for a mess-free workspace.