Learn More
Cayman Chemical 12-epI LeukotrIen B4 100ug
SDP
Catalog No. 502982970
Encompass_Preferred
An isomer of LTB4 which would not be expected to occur in either non-enzymatic hydrolysis products, or in the enzymatic products of LTA4 hydrolase; has significantly reduced activity compared to LTB4
Catalog No. 50-298-2970 Supplier Cayman Chemical Supplier No. 20135100 UG
An isomer of LTB4 which would not be expected to occur in either non-enzymatic hydrolysis products, or in the enzymatic products of LTA4 hydrolase; has significantly reduced activity compared to LTB4