Learn More
Cayman Chemical 12-epI LeukotrIen B4-d4 50ug
SDP
Catalog No. 502946170
Encompass_Preferred
An internal standard for the quantification of 12-epi LTB4 by GC- or LC-MS
Catalog No. 50-294-6170 Supplier Cayman Chemical Supplier No. 32013550 UG
An internal standard for the quantification of 12-epi LTB4 by GC- or LC-MS