Learn More
Cayman Chemical 12-EpInapelIn 10mg
SDP
Catalog No. 502913844
Encompass_Preferred
A diterpenoid alkaloid; has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia and antifibrillatory activity in a mouse model of aconitine-induced cardiac fibrillation (ED50s = 8 and 20 mg/kg, respectively)
Catalog No. 50-291-3844 Supplier Cayman Chemical Supplier No. 3655010 MG
A diterpenoid alkaloid; has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia and antifibrillatory activity in a mouse model of aconitine-induced cardiac fibrillation (ED50s = 8 and 20 mg/kg, respectively)