Learn More
Cayman Chemical 12-Epinapelline, Purity: ≥95%, CAS: 110064-71-6, 5 mg
SDP
Catalog No. 502913779
Encompass_Preferred
A diterpenoid alkaloid; has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia and antifibrillatory activity in a mouse model of aconitine-induced cardiac fibrillation (ED50s = 8 and 20 mg/kg, respectively)
Catalog No. 50-291-3779 Supplier Cayman Chemical Supplier No. 365505 MG
A diterpenoid alkaloid; has antiarrhythmic activity in a rat model of aconitine-induced arrhythmia and antifibrillatory activity in a mouse model of aconitine-induced cardiac fibrillation (ED50s = 8 and 20 mg/kg, respectively)