Learn More
Medchemexpress LLC 12-Epinapelline | 110064-71-6 | 5 MG
SDP
Catalog No. 5000420541
Encompass_Preferred
12-Epinapelline is a diterpene alkaloid isolated from Aconitum baikalense. It exhibits anti-inflammatory activity and stimulates the growth of colonies from fibroblast precursors. The initial source is identified as Aconitum carmichaeli Debx. from the Ranunculaceae family of plants.
  • Isolated from Aconitum baikalense
  • Exhibits anti-inflammatory activity
  • Stimulates the growth of colonies from fibroblast precursors
  • Derived from Aconitum carmichaeli Debx.
Catalog No. 50-004-20541 Supplier Medchemexpress LLC Supplier No. HYN21625MG
12-Epinapelline is a diterpene alkaloid isolated from Aconitum baikalense. It exhibits anti-inflammatory activity and stimulates the growth of colonies from fibroblast precursors. The initial source is identified as Aconitum carmichaeli Debx. from the Ranunculaceae family of plants.
  • Isolated from Aconitum baikalense
  • Exhibits anti-inflammatory activity
  • Stimulates the growth of colonies from fibroblast precursors
  • Derived from Aconitum carmichaeli Debx.