Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-epinapelline | 110064-71-6 | 99.9% | 359.50 g/mol | C22H33NO3 | 10 MG
SDP

Catalog No. 5000296185
Encompass_Preferred

12-epinapelline is a diterpene alkaloid isolated from Aconitum baikalense. It has demonstrated anti-inflammatory activity and promotes the growth of colonies from fibroblast precursors. The compound is a white to off-white solid with high purity and defined storage recommendations.

  • Exhibits anti-inflammatory activity
  • Stimulates fibroblast colony growth
  • Chemical formula C22H33NO3 and molecular weight 359.50 g/mol
  • White to off-white solid appearance
  • High purity (99.92% as listed by manufacturer)
  • Available in small research pack sizes suitable for biological assays
  • Store protected from light at 4°C; in solvent, long-term storage at -80°C

Catalog No. 50-002-96185 Supplier Medchemexpress LLC Supplier No. HYN216210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-epinapelline is a diterpene alkaloid isolated from Aconitum baikalense. It has demonstrated anti-inflammatory activity and promotes the growth of colonies from fibroblast precursors. The compound is a white to off-white solid with high purity and defined storage recommendations.

  • Exhibits anti-inflammatory activity
  • Stimulates fibroblast colony growth
  • Chemical formula C22H33NO3 and molecular weight 359.50 g/mol
  • White to off-white solid appearance
  • High purity (99.92% as listed by manufacturer)
  • Available in small research pack sizes suitable for biological assays
  • Store protected from light at 4°C; in solvent, long-term storage at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.