Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-Ethyl-9-hydroxycamptothecin | 119577-28-5 | 392.40 | 5 MG
SDP

Catalog No. 5000415228
Encompass_Preferred

12-Ethyl-9-hydroxycamptothecin is a derivative of Camptothecin, an alkaloid and a DNA topoisomerase I (Topo I) inhibitor. This compound is used in various research applications.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C22H20N2O5
  • Structure classification: Alkaloids, phenols
  • Initial source: Plants other families
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-004-15228 Supplier Medchemexpress LLC Supplier No. HYN20635MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Ethyl-9-hydroxycamptothecin is a derivative of Camptothecin, an alkaloid and a DNA topoisomerase I (Topo I) inhibitor. This compound is used in various research applications.

  • Purity: 99.89%
  • Appearance: Solid
  • Color: White to off-white
  • Formula: C22H20N2O5
  • Structure classification: Alkaloids, phenols
  • Initial source: Plants other families
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.