Learn More
Justrite™ 12 Gallons Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet With Manual Door
GREENER_CHOICE
Catalog No. 19822390 Shop All Justrite Safety Group Products
Change view
Click to view available options
Door Style:
Manual-close
Catalog No. Door Style
19-822-390 Manual-close
1 options
Catalog No. 19-822-390 Supplier Justrite™ Supplier No. 891202
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 12 Gallons Sure-Grip™ EX Compac Corrosives/Acid Steel Safety Cabinet With Manual Door is specially designed for safely storing acids and corrosives in laboratories.
Countertop and Compac Cabinets let you customize your work areas and make the most of available space. Whether adding to existing storage space or making chemicals readily available at workstation locations, these smaller-sized cabinets are the solution when space is at a premium.
Specifications
Dimensions (D x W x H) 18 x 23.25 x 35 in.
Description Cold-rolled Steel (18 ga)
Load Bearing Capacity 12 gal.
Color Blue
Material Steel (CR)
Door Style Manual-close