Learn More
MSC Empire Level Combination Square, 12 in., Stainless Steel, 1 ea
Catalog No. 502241064
Encompass
This combination square set offers precision machined working surfaces and a head with rational dimensions for quick checks of common measurements. It features a BLADE-LOCK™ self-aligning drawbolt for quick adjustments and a secure hold. The DUAL-PITCH™ vial allows for easy checks of level, one-, and two-degree slopes. The 12 in. etched stainless steel blade is designed not to fade over time and has high-visibility markings. The set also includes onboard storage for a hardened scriber.
  • Accurately measures distances and angles
  • Etched steel blade with heavy-duty machined square head
  • Hardened scriber marks surfaces
  • Self-aligning draw bolt for quick measurement changes
  • True Blue vial ensures accuracy
  • Warranty against manufacturer defects
This item is not returnable. View return policy
Catalog No. 50-224-1064 Supplier MSC Supplier No. 61927760
This combination square set offers precision machined working surfaces and a head with rational dimensions for quick checks of common measurements. It features a BLADE-LOCK™ self-aligning drawbolt for quick adjustments and a secure hold. The DUAL-PITCH™ vial allows for easy checks of level, one-, and two-degree slopes. The 12 in. etched stainless steel blade is designed not to fade over time and has high-visibility markings. The set also includes onboard storage for a hardened scriber.
  • Accurately measures distances and angles
  • Etched steel blade with heavy-duty machined square head
  • Hardened scriber marks surfaces
  • Self-aligning draw bolt for quick measurement changes
  • True Blue vial ensures accuracy
  • Warranty against manufacturer defects