Learn More
Cayman Chemical (±)12-HETE, 71030-37-0, 100 µg
SDP
Supplier:  Cayman Chemical 34550100
Encompass_Preferred
(±)12-HETE is one of the six monohydroxy fatty acids produced by the non-enzymatic oxidation of arachidonic acid. The biological activity of (±)12-HETE is similar to that of its constituent enantiomers (Item Nos. 34560 and 34570). It aggregates neutrophils with an EC50 value of 40 nM.{2091}
Catalog No. NC0714897