Learn More
SAI Infusion Technologies High Flow Extension Line - 12", Male & Female Luers
SDP
Catalog No. NC1632005
Encompass
12" High Flow Extension line with male and female luer adapters, 0.4ml dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 50/pk. Product is for laboratory animal research only.
Catalog No. NC1632005 Supplier SAI Infusion Technologies Supplier No. EXT12HF
12" High Flow Extension line with male and female luer adapters, 0.4ml dead volume. SAI extension lines are ideal for connecting between pump and swivel or pump and catheter in animal infusion studies. Made with non-DEHP PVC tubing and available in a variety of standard lengths. Custom sizes are also available. 50/pk. Product is for laboratory animal research only.