Learn More
Cayman Chemical 12-hydroxy Lauric Acid, Purity: ≥95%, CAS: 505-95-3, 5 g
SDP
Catalog No. 502940635
Encompass_Preferred
A hydroxylated fatty acid; active against various bacteria (MICs = 6.25-125 μg/ml) and the fungus C. albicans (MIC = 15.63 μg/ml)
Catalog No. 50-294-0635 Supplier Cayman Chemical Supplier No. 284945 G
A hydroxylated fatty acid; active against various bacteria (MICs = 6.25-125 μg/ml) and the fungus C. albicans (MIC = 15.63 μg/ml)