Learn More
Cayman Chemical 12-hydroxy Stearic Acid, Purity: ≥85%, CAS: 106-14-9, 1 g
SDP
Catalog No. 502947583
Encompass_Preferred
A hydroxy fatty acid; a low molecular weight gelator that self-assembles to form organogels; administration of paclitaxel in 12-hydroxy stearic acid-containing gel nanocarriers enhances tumor growth suppression in an H22 murine hepatocellular carcinoma model
Catalog No. 50-294-7583 Supplier Cayman Chemical Supplier No. 345581 G
A hydroxy fatty acid; a low molecular weight gelator that self-assembles to form organogels; administration of paclitaxel in 12-hydroxy stearic acid-containing gel nanocarriers enhances tumor growth suppression in an H22 murine hepatocellular carcinoma model