Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-Hydroxydodecanoic acid | 505-95-3 | 98.9% | 1 ML
SDP

Catalog No. 5000342905
Encompass_Preferred

12-Hydroxydodecanoic acid is an endogenous metabolite provided as a 10 mM solution in 1 mL of DMSO. This high-purity compound is intended for research use.

  • Endogenous metabolite
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • Purity of 98.87%
  • Molecular weight: 216.32
  • Chemical formula: C12H24O3

Catalog No. 50-003-42905 Supplier Medchemexpress LLC Supplier No. HY12874310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Hydroxydodecanoic acid is an endogenous metabolite provided as a 10 mM solution in 1 mL of DMSO. This high-purity compound is intended for research use.

  • Endogenous metabolite
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • Purity of 98.87%
  • Molecular weight: 216.32
  • Chemical formula: C12H24O3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.