Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-Hydroxydodecanoic acid | 505-95-3 | 98.9% | 100 MG
SDP

Catalog No. 5000340934
Encompass_Preferred

12-Hydroxydodecanoic acid is an endogenous metabolite, presenting as a white to off-white solid. It is classified under ketones, aldehydes, and acids, and has been tested for agonist activity at recombinant human GPR84 expressed in CHO cell membranes.

  • Endogenous metabolite
  • Molecular weight: 216.32
  • Chemical formula: C12H24O3
  • White to off-white solid appearance
  • Classified as a ketone, aldehyde, and acid
  • Shows EC50 > 100 μM for agonist activity at recombinant human GPR84 in CHO cell membranes

Catalog No. 50-003-40934 Supplier Medchemexpress LLC Supplier No. HY128743100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Hydroxydodecanoic acid is an endogenous metabolite, presenting as a white to off-white solid. It is classified under ketones, aldehydes, and acids, and has been tested for agonist activity at recombinant human GPR84 expressed in CHO cell membranes.

  • Endogenous metabolite
  • Molecular weight: 216.32
  • Chemical formula: C12H24O3
  • White to off-white solid appearance
  • Classified as a ketone, aldehyde, and acid
  • Shows EC50 > 100 μM for agonist activity at recombinant human GPR84 in CHO cell membranes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.