Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-Hydroxydodecanoic acid | 505-95-3 | 98.9% | 50 MG
SDP

Catalog No. 5000342872
Encompass_Preferred

12-Hydroxydodecanoic acid is an endogenous metabolite.

  • For research use only.
  • Not sold to patients.
  • Appears as a white to off-white solid.
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month.
  • Solubility in DMSO: 100 mg/mL (462.28 mM) (requires ultrasonic, use newly opened DMSO).
  • This product is a controlled substance and may not be for sale in all territories.

Catalog No. 50-003-42872 Supplier Medchemexpress LLC Supplier No. HY12874350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Hydroxydodecanoic acid is an endogenous metabolite.

  • For research use only.
  • Not sold to patients.
  • Appears as a white to off-white solid.
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; In solvent at -80°C for 6 months, -20°C for 1 month.
  • Solubility in DMSO: 100 mg/mL (462.28 mM) (requires ultrasonic, use newly opened DMSO).
  • This product is a controlled substance and may not be for sale in all territories.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.