Learn More
Grainger 12 In Converstion Kit Steel 20 In Dia
Catalog No. 502768068
Encompass_Preferred
The Grainger 16Y907 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" x 1/2" hand pad has an open web construction with abrasive particles throughout for scrubbing and scouring. It is made of high-quality fibers and minerals that resist shedding. The pad can be used wet or dry on surfaces including stainless steel, porcelain, fiberglass and painted wood. It effectively removes grease, dirt, food residue and more. The 3M Scotch-Brite General Purpose Hand Pad provides versatile cleaning while being gentle on surfaces.
Catalog No. 50-276-8068 Supplier Grainger Supplier No. 16Y907
The Grainger 16Y907 is a 3M Scotch-Brite General Purpose Hand Pad that can be used for a variety of cleaning applications. This 8" x 4" x 1/2" hand pad has an open web construction with abrasive particles throughout for scrubbing and scouring. It is made of high-quality fibers and minerals that resist shedding. The pad can be used wet or dry on surfaces including stainless steel, porcelain, fiberglass and painted wood. It effectively removes grease, dirt, food residue and more. The 3M Scotch-Brite General Purpose Hand Pad provides versatile cleaning while being gentle on surfaces.