Learn More
Cayman Chemical 12-keto Lithocholic Acid, Purity: ≥98%, CAS: 5130-29-0, 50 mg
SDP
Catalog No. 502989438
Encompass_Preferred
A metabolite of DCA; fecal levels are elevated in rats fed a diet supplemented with cholic acid or DCA compared with rats fed a control diet or a diet supplemented with cholesterol, HDCA, or LCA; activates the human PXR (EC50 = 31.3 μM in a cell-based transactivation assay); levels are increased in duodenal bile from patients with type 2 diabetes
Catalog No. 50-298-9438 Supplier Cayman Chemical Supplier No. 2954750 MG
A metabolite of DCA; fecal levels are elevated in rats fed a diet supplemented with cholic acid or DCA compared with rats fed a control diet or a diet supplemented with cholesterol, HDCA, or LCA; activates the human PXR (EC50 = 31.3 μM in a cell-based transactivation assay); levels are increased in duodenal bile from patients with type 2 diabetes