Learn More
Cayman Chemical 12-keto LIthocholIc AcId 5mg
SDP
Catalog No. 502992645
Encompass_Preferred
A metabolite of DCA; fecal levels are elevated in rats fed a diet supplemented with cholic acid or DCA compared with rats fed a control diet or a diet supplemented with cholesterol, HDCA, or LCA; activates the human PXR (EC50 = 31.3 μM in a cell-based transactivation assay); levels are increased in duodenal bile from patients with type 2 diabetes
Catalog No. 50-299-2645 Supplier Cayman Chemical Supplier No. 295475 MG
A metabolite of DCA; fecal levels are elevated in rats fed a diet supplemented with cholic acid or DCA compared with rats fed a control diet or a diet supplemented with cholesterol, HDCA, or LCA; activates the human PXR (EC50 = 31.3 μM in a cell-based transactivation assay); levels are increased in duodenal bile from patients with type 2 diabetes