Learn More
Apexbio Technology LLC 12-KETOLITHOCHOLIC-ACID-1MG
SDP
Catalog No. 5000567880
Encompass
12-Ketolithocholic acid (CAS No 5130-29-0) is a bile acid and a metabolite of lithocholic acid studied within the biomedical research category for its role in hepatobiliary diseases and cholesterol metabolism Originating from the liver s enzymatic processes it interacts with nuclear receptors such as FXR impacting the expression of genes involved in bile acid homeostasis and lipid metabolism In vitro it exhibits activity in the nanomolar to low micromolar range influencing pathways relevant to cholesterol regulation Its applications include cell model experiments and animal studies focused on liver function and metabolic disorders Additionally it serves a role in drug screening for potential therapeutic agents targeting liver diseases Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67880 Supplier Apexbio Technology LLC Supplier No. C86061
Add to Cart
Add to Cart
12-Ketolithocholic acid (CAS No 5130-29-0) is a bile acid and a metabolite of lithocholic acid studied within the biomedical research category for its role in hepatobiliary diseases and cholesterol metabolism Originating from the liver s enzymatic processes it interacts with nuclear receptors such as FXR impacting the expression of genes involved in bile acid homeostasis and lipid metabolism In vitro it exhibits activity in the nanomolar to low micromolar range influencing pathways relevant to cholesterol regulation Its applications include cell model experiments and animal studies focused on liver function and metabolic disorders Additionally it serves a role in drug screening for potential therapeutic agents targeting liver diseases Experimental concentrations typically depend on the specific research design