Learn More
Ace Glass, Inc. Flask, freeze drying, 1.2L, 249mm height, 53mm ID neck, 100mm body OD, includes snap-on rubber cap and 1/2in OD PTFE valved manifold connection adapter
SDP
Catalog No. 5097434
Encompass_Preferred
Catalog No. 50-974-34 Supplier Ace Glass, Inc. Supplier No. 703556