Learn More
Cayman Chemical 12-Lipoxygenase leukocyte-ty
SDP
Catalog No. 502947283
Encompass_Preferred
Immunogen: Full length recombinant mouse 12-lipoxygenase (leukocyte-type) • Host: Rabbit • Species Reactivity: (+) Mouse, porcine • Applications: ELISA, WB • MW = ~73 kDa
Catalog No. 50-294-7283 Supplier Cayman Chemical Supplier No. 32822500 UG
Add to Cart
Add to Cart
Immunogen: Full length recombinant mouse 12-lipoxygenase (leukocyte-type) • Host: Rabbit • Species Reactivity: (+) Mouse, porcine • Applications: ELISA, WB • MW = ~73 kDa