Learn More
AirSea USA 12L UN Steel Drum, 1A2/Y1.8/200 & 1A2/X28/S
Supplier:  AirSea USA 0437
Encompass_Preferred
UN approved packaging - 1A2/1.8/200 & 1A2/X28/S 12L UN steel drum UN Marks: 1A2/Y1.8/200 & 1A2/X28/S 12L UN Steel Drum with Lacquer Lining & Lever Locking Ring - approved to ship both solid and liquid dangerous goods substances.
Catalog No. NC9025959