Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Mercaptododecylphosphonic acid, 95%

Catalog No. AAH3396903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3396903 1 g
AAH33969MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3396903 Supplier Thermo Scientific Chemicals Supplier No. H3396903
Only null left
Add to Cart

CAS: 159239-33-5 | C12H27O3PS | 282.38 g/mol

Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159239-33-5
Molecular Formula C12H27O3PS
Molecular Weight (g/mol) 282.38
MDL Number MFCD11046116
InChI Key PVIUMTORLYKRJT-UHFFFAOYSA-N
Synonym 12-mercaptododecylphosphonic acid, 12-sulfanyldodecyl phosphonic acid
PubChem CID 45158994
IUPAC Name 12-sulfanyldodecylphosphonic acid
SMILES OP(O)(=O)CCCCCCCCCCCCS

Specifications

Melting Point 94°C to 96°C
Linear Formula HS(CH2)12P(O)(OH)2
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 282.38
Percent Purity 95%
Chemical Name or Material 12-Mercaptododecylphosphonic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.