Learn More
Enzo Life Sciences 12-Methoxydodecanoic acid (50mg). CAS: 92169-28-3
SDP
Catalog No. 502010866
Encompass_Preferred
A myristate analog which has similar chain length but reduced hydrophobicity compared to myristate. Inhibits replication of HIV-1. Its specific mechanism of action is not clear but it is believed to either inhibit the activity of N-myristoyltransferase or modulate the function of target acylated proteins. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25 mg/mL) and DMSO (25 mg/mL). Long Term Storage: Ambient.
Catalog No. 50-201-0866 Supplier Enzo Life Sciences Supplier No. BMLG2340050
A myristate analog which has similar chain length but reduced hydrophobicity compared to myristate. Inhibits replication of HIV-1. Its specific mechanism of action is not clear but it is believed to either inhibit the activity of N-myristoyltransferase or modulate the function of target acylated proteins. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (25 mg/mL) and DMSO (25 mg/mL). Long Term Storage: Ambient.