Learn More
Cayman Chemical 12-methyl Myristic Acid, Purity: ≥95%, CAS: 5502-94-3, 1 mg
SDP
Catalog No. 502932859
Encompass_Preferred
A methylated fatty acid; has been found in bacteria, dairy products, wild-caught and farm-raised saltwater fish, and mouse subcutaneous adipose tissue; reduces larval settlement of H. elegans in a concentration-dependent manner (0.625-40 μg/ml); levels are decreased in mice fed a high-fat diet
Catalog No. 50-293-2859 Supplier Cayman Chemical Supplier No. 248171 MG
A methylated fatty acid; has been found in bacteria, dairy products, wild-caught and farm-raised saltwater fish, and mouse subcutaneous adipose tissue; reduces larval settlement of H. elegans in a concentration-dependent manner (0.625-40 μg/ml); levels are decreased in mice fed a high-fat diet